Learn More
Thermo Scientific™ HAAKE™ RheoWin™ Measuring and Evaluation Software
Product Code. 17916850
Change view
Click to view available options
Type:
Software
Software Module
For Use With (Equipment):
HAAKE MARS 40 Rheometer
HAAKE MARS 60 Rheometer
HAAKE MARS iQ Air Rheometer
HAAKE RheoWin Software
HAAKE Viscotester iQ
HAAKE Viscotester iQ Air
For Use With (Application):
21 CFR part 11 tool
Navigating System Operations
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type For Use With (Equipment) For Use With (Application) Unit Size
17916850 Software HAAKE MARS 40 Rheometer Navigating System Operations Each
15375123 Software HAAKE Viscotester iQ Navigating System Operations Each
15528269 Software HAAKE Viscotester iQ Air Navigating System Operations -
17387199 Software HAAKE MARS iQ Air Rheometer Navigating System Operations Each
17630687 Software HAAKE MARS 60 Rheometer Navigating System Operations Each
17906850 Software Module HAAKE RheoWin Software 21 CFR part 11 tool Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 17916850 Supplier Thermo Scientific™ Supplier No. 0985072
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
This item is not returnable. View return policy
Easily navigate your rheometer or viscometer operation using RheoWin Measuring and Evaluation Software.
Easily navigate your rheometer or viscometer operation using the Thermo Scientific™ HAAKE™ RheoWin™ Measuring and Evaluation Software. This user-friendly software for rheometers and viscometers can be customized to offer convenient rheometer or viscometer operation with default settings for beginners and full access to all relevant measuring parameters for advanced rheologists.
The HAAKE RheoWin Measuring and Evaluation Software Features
Customizable Software
  • RheoWin JobManager for fully automated process control ('jobs') of measuring and analysis routines and report printout or export
  • RheoWin DataManager for interactive evaluation of measured data, as well as sophisticated tools for creating reports and generating templates for graphs, tables and screen views
  • RheoWin UserManager for comprehensive management of user access control and assignment of specific access rights
Functionality
  • Manual control for preliminary testing, selected parameters display and saving manually-acquired data
  • Convenient creation and customization of measuring routines using predefined measuring and evaluation elements via 'drag and drop' techniques
  • Fully automated measuring, data evaluation and documentation within one measuring routine
  • Simultaneous measurements using several instruments and data evaluation
  • Freely configurable data export (e.g., ASCII, MS-Excel, XML, etc.)
  • Ability to save graphs in various formats
  • Numerous algorithms for data analysis (e.g., interpolation, regression and automated quality control)
  • Ability to save raw data and numerical values for quality control purposes
  • Integrated image capture with USB and firewire camera
Customization
  • User-defined configuration of paths and subdirectories for data filing
  • Modular generation of a file name and automated saving in a predefined subdirectory
  • Data transfer to ERP and laboratory systems (e.g., SAP™, LIMS, etc.)
  • Online display of all corrections to improve data quality
  • Snapshot for quick characterization of an unknown sample
Operating System:
Windows 10 (32-bit and 64-bit version)
Windows 8.1 (32-bit and 64-bit version)
Windows 8 (32-bit and 64-bit version)
Windows 7 (32-bit and 64-bit version)
TRUSTED_SUSTAINABILITY
Specifications
Type Software
For Use With (Equipment) HAAKE MARS 40 Rheometer
For Use With (Application) Navigating System Operations