Learn More
Hanna™ Instruments Standard DO Probe
Product Code. 10469880
Change view
Click to view available options
Length (Metric) Cable:
4 m
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Cable Unit Size
10469880 4 m Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10469880 Supplier Hanna Instruments Supplier No. HI76407A/P
  • ONLINE EXCLUSIVE PRICE
    275.09€
    136.76€ / 5 pièces
    Économisez 138.33€
    50% de réduction
Expédition estimée: 22-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Type Dissolved Oxygen Screw Cap Membranes
Length (Metric) Cable 4 m
For Use With (Equipment) Hanna portable polarographic dissolved oxygen probes
Membrane Polytetrafluoroethylene (PTFE)