Learn More
Sampling Systems™ SteriWare™ HDPE Disposable Powder Sampler
BIOPHARMA
Product Code. 15200749 Shop All Sampling Systems Products
Change view
Click to view available options
Capacity (Metric):
100 mL
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15200749 100 mL Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15200749 Supplier Sampling Systems™ Supplier No. 8240H1000S
  • ONLINE EXCLUSIVE PRICE
    399.00€
    355.13€ / 20 pièces
    Économisez 43.87€
    10% de réduction
View all promotions
Expédition estimée: 15-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Can be used straight from the bag. The SteriWare™ Powder Sampler is ideal for taking large volume samples from powders or granules.
Key Features:
  • Assembled and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming HDPE
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • Suitable for use in ATEX areas
Operation:
  • Push the sampler into the product. Ensuring he sampler is in the closed position.
  • Rotate handle to open up the slot.
  • After leaving the sampler for several seconds to allow powder to enter, rotate handle to close the sampler.
  • Remove sampler from the product.
  • Deposit the sample into a suitable container.
Why Use a SteriWare™ Disposable Sampler?
  • No Cross Contamination - All SteriWare™ devices can be used straight from the bag.
  • No Cleaning - Unlike a reusable sampler, you can use a SteriWare™ product straight away. This eliminates the need for costly Cleaning Validation.
  • High Quality - Our SteriWare™ products are manufactured/moulded/packed in a Class 100,000 Cleanroom in England. Please see the products Certificate of Conformity and Technical Specification Sheet for more information.
  • Full Documentation - We can provide full batch specific documentation for all SteriWare™ products. This includes Certificates of Conformity and Certificates of Irradiation (where applicable). The batch number is clearly printed on every bag and box.
  • Cost Effective - Removing the need for Cleaning Validation by switching to a SteriWare™ device saves time and money.
  • Available Sterile - Most SteriWare™ products are available pre-sterilized by Gamma or Electron Beam to SAL10-6. Please see the Specification Sheet for more information.
Specifications
Product Type Powder Sampler
Material HDPE
Capacity (Metric) 100 mL
Disposable Single-use
Sterility Sterile
Certifications/Compliance Conforms to FDA CFR 177.1520, EU Regulations 10/2011, EU Regulations 10/2011
For Use With (Application) For taking a large volume sample from a single known point
Length (Metric) 987 mm