Learn More
WHEATON™ WHEATON™ HDPE Liquid Scintillation Vial
Product Code. 11758869 Shop All DWK Life Sciences Products
Change view
Click to view available options
Material (Liner):
Foamed Polyethylene
Linerless
Metal foil
Quantity:
1000/Pk.
500/Pk.
Unit Size:
Pack of 1000
Pack of 500
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material (Liner) Quantity Unit Size
11758869 Foamed Polyethylene 1000/Pk. Pack of 1000
11754179 Linerless 1000/Pk. Pack of 1000
10523804 Linerless 500/Pk. Pack of 500
11748839 Metal foil 1000/Pk. Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11758869 Supplier WHEATON™ Supplier No. 986720
  • ONLINE EXCLUSIVE PRICE
    436.00€
    364.15€ / 1000 pièces
    Économisez 71.85€
    16% de réduction
Expédition estimée: 25-02-2027
Add to basket
Add to basket
This item is not returnable. View return policy
WHEATON™ HDPE Liquid Scintillation Vial is designed for reliable liquid scintillation counting. Made from high-density polyethylene with lightweight walls for increased counting efficiency, manufactured to precise tolerances, and available with a choice of cap liner materials.
  • Made from high-density polyethylene (HDPE) with lightweight walls for increased counting efficiency
  • Manufactured to precise tolerances to avoid jamming in automated equipment
  • 20 mL capacity with 22-400 cap size; dimensions 27 x 61 mm
  • Available with a choice of cap liner materials including linerless, foamed polyethylene, and metal foil options
  • Cases of 1000 vials are packed in a single plastic bag with caps supplied separately
  • Suitable for buffers, reagents, and liquid, solid, and semi-solid content
Specifications
Material (Liner) Foamed Polyethylene
Color Natural
Diameter (Metric) Outer 27 mm
Material HDPE
Sterility Non-sterile
Volume (Metric) 20 mL
Closure Material Polypropylene
Closure Type Screw Cap
Quantity 1000/Pk.
Closure Size 22-400
Autoclavable Not Autoclavable
Height (Metric) 61 mm
Product Type Scintillation Vial