Learn More
Labconco™ Heated Shelf with Sensor, Bulk Tray Dryer
Product Code. 13484890 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Labconco™ FreeZone™ Bulk Tray Dryers
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
13484890 Labconco™ FreeZone™ Bulk Tray Dryers Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13484890 Supplier Labconco™ Supplier No. 7760801
  • ONLINE EXCLUSIVE PRICE
    1250.00€
    1110.21€ / 1 pièces
    Économisez 139.79€
    11% de réduction
 Restricted Item
Expédition estimée: 24-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
The Heated Shelf with Sensor is for use with Labconco™ FreeZone™ Bulk Tray Dryers.
  • 200-watt heater ensures efficient heating from −20°C to 60°C.
  • A microprocessor in the Bulk Tray Dryer maintains system temperature within 3°C of set point.
  • FreeZone™ Bulk Tray Dryers include three and can accommodate two additional Heated Shelves with Sensors.
Specifications
Type Shelf
For Use With (Equipment) Labconco™ FreeZone™ Bulk Tray Dryers
Depth (English) 16.6 in.
Depth (Metric) 42.2 cm
Width (English) 12.7 in.
Width (Metric) 32.1 cm
Voltage 230 V
Frequency 50 Hz
Shipping Weight (English) 5 lb.
Shipping Weight (Metric) 2.3 kg