Learn More
Thermo Scientific™ HeSaver-H2Safer™ kit for iConnect™ SSL injector module TRACE™ 1600 Series GC
Product Code. 18990080 Shop All Thermo Scientific Products
Change view
Click to view available options
Injection Mode:
Split/Splitless
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Injection Mode Unit Size
18990080 Split/Splitless Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18990080 Supplier Thermo Scientific™ Supplier No. MI1900000022
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
This item is not returnable. View return policy
Easily convert your iConnect SSL to save significantly on carrier gas consumption, whether using helium or hydrogen, and maintain your current GC methods.
Improve the sustainability of your laboratory, regardless of whether helium or hydrogen is your carrier gas. To significantly reduce carrier gas consumption while running samples and when the GC is idle, the Thermo Scientific HeSaver-H2Safer kit easily transforms the iConnect SSL injector module. To save time and cost for re-validation, existing methods remain unaffected. When helium is used as carrier, the HeSaver mode substantially extends cylinder lifetime and mitigates helium shortage concerns. When hydrogen is used as carrier, the H2Safer mode ensures safer operations and optimized use of hydrogen generators.
Maintain your existing methods and dramatically save on carrier gas consumption, whether using helium or hydrogen carrier gas:
For laboratories using helium as carrier gas
  • Mitigate helium shortage concerns, including delayed delivery and increased costs
  • Maintain existing validated methods
  • Achieve optimal GCMS performance with no compromise on sensitivity or spectra matching
For laboratories using hydrogen as carrier gas
  • Benefit from safer operating conditions thanks to a limited max flow rate, removing the need for hydrogen sensor in the GC oven
  • Serve more GCs with a hydrogen generator
  • Ensure inert injection environment in the hot SSL since hydrogen is not flushing into the injector
Additional benefits of using the HeSaver-H2Safer option
  • Save time for inlet maintenance. The modified SSL injector can be accessed while the carrier gas flows into the column, removing the need to cool the GC oven.
  • Reduce contamination from the injector into the column. The carrier gas does not flow through the injector, which protects the column from common contaminants (septum/sample matrix/by-products).
TRUSTED_SUSTAINABILITY
Specifications
Product Type Kit for iConnect SSL injector module
Injection Mode Split/Splitless
Description HeSaver-H2Safer™ kit for iConnect™ SSL injector module TRACE™ 1600 Series GC
For Use With (Equipment) iConnect SSL injector on TRACE 1600 Series GC. Compatible with capillary columns from 50 µm to 320 µm inner diameter
Quantity Each