Learn More
LNI Swissgas HG 2U PRO 100
Product Code. 17882219 Shop All LNI Swissgas Products
Change view
Click to view available options
Voltage:
100/115/220/230 V
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Voltage Unit Size
17882219 100/115/220/230 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 17882219 Supplier LNI Swissgas Supplier No. 6920.16.010
  • ONLINE EXCLUSIVE PRICE
    9465.00€
    8967.20€ / 1 pièces
    Économisez 497.80€
    5% de réduction
Expédition estimée: 21-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Hydrogen generator ultra high purity.
  • Accessory
Specifications
Voltage 100/115/220/230 V
Type Hydrogen Carrier Gas Generator