Learn More
Nabertherm™ High-Temperature Ovens, Forced Convection Chamber Furnaces
Product Code. 17398049 Shop All Nabertherm Products
Change view
Click to view available options
Capacity (Metric):
30 L
60 L
120 L
250 L
500 L
Controller Type:
B400
B500
B510
C540
C550
Unit Size:
Each
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Controller Type Unit Size
17398049 120 L B500 Each
17338049 60 L B500 Each
17348049 60 L C540 Each
17308059 120 L C540 Each
17358059 250 L B400 Each
17368059 250 L C540 Each
17318069 500 L B400 Each
17328069 500 L C540 Each
17378069 30 L B510 Each
17388069 30 L C550 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 17398049 Supplier Nabertherm™ Supplier No. NA1283K2
 Restricted Item
Expédition estimée: 03-02-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Chamber furnaces with air circulation are characterized by their extremely high temperature uniformity. Suitable for processes such as annealing, crystalizing, preheating, curing, and tool making.
  • Horizontal air circulation with optimum distribution through stainless steel baffles
  • Swing door hinged on the right
  • Temperature uniformity up to +/- 4 C according to DIN 17052-1 (NAT 15/65 and NAT 30/85 up to +/- 5°C) in the empty work space
  • One frame sheet and rails for two additional trays included in the scope of delivery (NAT 15/65 and NAT 30/85 without frame sheet)
  • Base frame included in the delivery, NAT 15/65 and NAT 30/85 designed as table-top model
Please Note: A furnace below 2.9 kW will be delivered with a standard UK plug. Anything above 2.9 kW will be delivered with the appropriate CEE plug. For anything above 2.9 kW, please ensure you have the required socket prior to purchase.
Specifications
Controller Type B500
Capacity (Metric) 120 L
Temperature (Metric) 850°C
Dimensions (D x W x H) Interior 600 x 450 x 450 mm
Dimensions (L x W x H) Exterior 1420 x 890 x 1540 mm
Voltage 400 V
Wattage 14000 W