Learn More
Hoefer™ Vacuum Gel Dryer
Product Code. 12855876
Change view
Click to view available options
Voltage:
230 V
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Voltage Unit Size
12855876 230 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12855876 Supplier Hoefer Supplier No. GD2002
Expédition estimée: 16-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
The user can set independent timers for heat and for vacuum
The user can set independent timers for heat and for vacuum. The thermostat is adjustable from 40°C to 80°C, ±2°C - temperatures that vaporise water but do not burn the gels
  • Dries acrylamide and agarose gels up to 330mm x 440mm in as little as 30min
  • The heavy aluminum platen provides even heat distribution and even vacuum distribution under the gel
  • A clear silicone cover sheet allows the user to view the gel during drying
  • The quiet, oil-free diaphragm pump maintains the low vacuum necessary for rapid gel drying. Requires no oil and no trap
Dry gels rapidly, evenly and safely, using a dry-heat vacuum method.
Specifications
Type Vacuum Gel Dryer
Voltage 230 V
Frequency 50/60 Hz