Learn More
Klever Holder for all Klever Cutters
Product Code. 17738997
Change view
Click to view available options
Color:
Black
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17738997 Black Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17738997 Supplier Klever Supplier No. KCJHOL
  • ONLINE EXCLUSIVE PRICE
    148.04€
    127.69€ / 10 pièces
    Économisez 20.35€
    13% de réduction
Expédition estimée: 24-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Klever Holster safely stores cutters and replacement heads, and conveniently Velcro onto your belt.
  • Stores cutter on the belt.
  • Also has a storage pocket for spare Xchange blades.
  • Durably constructed for years of reliable use.
Specifications
For Use With (Equipment) Klever Cutter Range
Length (English) 6.88 in.
Length (Metric) 17.5 cm
Color Black
Product Type Tool Holster