Learn More
Systec™ Horizontal, Front-Loading, Benchtop Autoclave, DE Series
Product Code. 12082959
Change view
Click to view available options
Capacity (Metric):
113 L (Total), 100 L (Nominal)
211 L (Total), 200 L (Nominal)
50 L (Total), 45 L (Nominal)
95 L (Total), 90 L (Nominal)
Depth (Metric) Chamber:
500 mm
700 mm
1000 mm
Diameter (Metric) Chamber:
344 mm
400 mm
500 mm
Model:
DE-100
DE-45
DE-90
DE200
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Depth (Metric) Chamber Diameter (Metric) Chamber Model Unit Size
12082959 113 L (Total), 100 L (Nominal) 500 mm 500 mm DE-100 Each
10405373 50 L (Total), 45 L (Nominal) 500 mm 344 mm DE-45 Each
12072959 95 L (Total), 90 L (Nominal) 700 mm 400 mm DE-90 Each
12002969 211 L (Total), 200 L (Nominal) 1000 mm 500 mm DE200 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Pour r\u00E9pondre \u00E0 vos besoins de st\u00E9rilisation, l'autoclave peut n\u00E9cessiter certaines options. Il est indispensable de remplir le guide de choix que vous retrouverez dans la section services / guides de choix
Regulatory: This product is under eco-contribution tax.
Product Code. 12082959 Supplier Systec™ Supplier No. 1600
  • ONLINE EXCLUSIVE PRICE
    19176.35€
    17955.64€ / 1 pièces
    Économisez 1220.71€
    6% de réduction
View all promotions
 Restricted Item
Expédition estimée: 29-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Pour r\u00E9pondre \u00E0 vos besoins de st\u00E9rilisation, l'autoclave peut n\u00E9cessiter certaines options. Il est indispensable de remplir le guide de choix que vous retrouverez dans la section services / guides de choix
Regulatory: This product is under eco-contribution tax.
Systec™ Horizontal, Front-Loading, Benchtop Autoclave, DE Series is economical, suitable for basic sterilization applications, and can accept limited additional process optimization options.
  • Standard features:
    • Internal heating elements within the autoclave chamber
    • Housing, support frame and pressure vessel made of corrosion-resistant stainless steel
    • Temperature and pressure range 140°C, 4 bar (except for Systec DE-23)
    • LCD display and fully automatic microprocessor control
    • Number of sterilization programs: 12
    • Code-secured access rights for changing parameters and further safety-relevant intervention
    • Internal memory for storing up to 500 sterilization cycles
    • Timer for starting programs
    • Autofill: automatic demineralized water feed for steam generation
    • Flexible PT-100 temperature sensor
    • Temperature holding function for liquids after program finish
    • Special program for Durham tubes
    • Calculation of F0 value
    • Special program for waste sterilization with pulsed heat-up for more efficient air exhaust
    • Water-cooled steam exhaust, thermostatically controlled
    • Programmable automatic door-opening on completion of program
    • RS-232 and RS-485 interfaces for external data transmission (network-compatible)
  • Additional options for temperature, pressure, cooling, exhaust filtration, and documentation are available.
Specifications
Capacity (Metric) 113 L (Total), 100 L (Nominal)
Depth (Metric) Chamber 500 mm
Diameter (Metric) Chamber 500 mm
Model DE-100
Product Type Horizontal, Front-loading Benchtop Autoclave