Learn More
SCAT™ HPLC Supply and Waste Set, V2.0, Suitable for All HPLC Systems
Product Code. 18636478 Shop All SCAT Europe Products
Change view
Click to view available options
Includes:
4 x Safety Cap III, GL45, 4 x air valve, 1 x Safety Waste Cap, S60/61, electrostatic conductive, 1 x Exhaust filter M, V3.0, with splash guard and change indicator, 1 x 5 liter canister, PE-HD-EL, 1 x Grounding cable, 2 x clamp, 1.5 m length, 4 x Laboratory bottle DURAN, GL45, 1000 mL, clear glass and round, with cap and pouring ring, 5 x PFA-fitting with integrated ferrule, 2.3 mm OD, violet, 5 x PFA fitting with integrated ferrule, 1.6 mm OD, green, 10 x PFA fitting with integrated ferrule, 3.2 mm OD, blue, 10 x PFA blind plug, 2 x connector, straight, 6 to 8 mm ID, 1 x connector, curved, 5 to 11.5 mm ID, 1 x connector, angled, 9.5 to 10 mm ID
4 x Safety Cap III, GL45, 8 x air valve, 1 x Safety Waste Cap, GL45, 1 x Exhaust filter L, V3.0, with splash guard and change indicator, service life 12 months, 1 x 5 litre canister, PE-HD, 4 x Laboratory bottle DURAN, GL45, 1000 ml, clear glass and round, with cap and pouring ring , 5 x PFA-fitting with integrated ferrule, 2.3 mm OD, violet, 5 x PFA fitting with integrated ferrule, 1.6 mm OD, green, 10 x PFA fitting with integrated ferrule, 3.2 mm OD, blue, 10 x PFA blind plug, 2 x connector, straight, 6 - 8 mm ID, 1 x connector, curved, 5.0 - 11.5mm ID, 1 x connector, angled, 9.5 - 10 mm ID
4 x Safety Cap III, GL45, 8 x air valve, 1 x Safety Waste Cap, S60/61, electrostatic conductive, 1 x Exhaust filter L, V3.0, with splash guard and change indicator, service life 12 month, 1 x 5 litre canister, PE-HD-EL, 1 x Grounding cable, 2x clamp, L = 1,5 m, 4 x Laboratory bottle DURAN, GL45, 1000 ml, clear glass and round, with cap and pouring ring , 5 x PFA-fitting with integrated ferrule, 2.3 mm OD, violet, 5 x PFA fitting with integrated ferrule, 1.6 mm OD, green, 10 x PFA fitting with integrated ferrule, 3.2 mm OD, blue, 10 x PFA blind plug, 2 x connector, straight, 6 - 8 mm ID, 1 x connector, curved, 5.0 - 11.5mm ID, 1 x connector, angled, 9.5 - 10 mm ID
4 x Safety Cap III, V2.0, GL45, 1 x Safety Waste Cap, V2.0, GL45, 1 x Exhaust filter M, V2.0, 4 x Laboratory bottle DURAN, GL45, 1000 mL, 4 x air valve, 5 x PFA fitting with integrated ferrule, 2.3 mm, violet, 5 x PFA fitting with integrated ferrule, 1.6 mm, green, 10 x PFA fitting with integrated ferrule, 3.2 mm, blue, 10 x Blanking plugs, 1.6/2.3/3.2 mm, colorless, 2 x Tube connector, straight 6 - 8 mm, 1 x Tube connector, curved, 5.0 - 11.5 mm, 1 x tube connector, angled, 9.5 - 10 mm
Unit Size:
1 set
1 sheet
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
18636478 4 x Safety Cap III, GL45, 8 x air valve, 1 x Safety Waste Cap, S60/61, electrostatic conductive, 1 x Exhaust filter L, V3.0, with splash guard and change indicator, service life 12 month, 1 x 5 litre canister, PE-HD-EL, 1 x Grounding cable, 2x clamp, L = 1,5 m, 4 x Laboratory bottle DURAN, GL45, 1000 ml, clear glass and round, with cap and pouring ring , 5 x PFA-fitting with integrated ferrule, 2.3 mm OD, violet, 5 x PFA fitting with integrated ferrule, 1.6 mm OD, green, 10 x PFA fitting with integrated ferrule, 3.2 mm OD, blue, 10 x PFA blind plug, 2 x connector, straight, 6 - 8 mm ID, 1 x connector, curved, 5.0 - 11.5mm ID, 1 x connector, angled, 9.5 - 10 mm ID 1 sheet
16458190 4 x Safety Cap III, V2.0, GL45, 1 x Safety Waste Cap, V2.0, GL45, 1 x Exhaust filter M, V2.0, 4 x Laboratory bottle DURAN, GL45, 1000 mL, 4 x air valve, 5 x PFA fitting with integrated ferrule, 2.3 mm, violet, 5 x PFA fitting with integrated ferrule, 1.6 mm, green, 10 x PFA fitting with integrated ferrule, 3.2 mm, blue, 10 x Blanking plugs, 1.6/2.3/3.2 mm, colorless, 2 x Tube connector, straight 6 - 8 mm, 1 x Tube connector, curved, 5.0 - 11.5 mm, 1 x tube connector, angled, 9.5 - 10 mm Each
17865704 4 x Safety Cap III, GL45, 4 x air valve, 1 x Safety Waste Cap, S60/61, electrostatic conductive, 1 x Exhaust filter M, V3.0, with splash guard and change indicator, 1 x 5 liter canister, PE-HD-EL, 1 x Grounding cable, 2 x clamp, 1.5 m length, 4 x Laboratory bottle DURAN, GL45, 1000 mL, clear glass and round, with cap and pouring ring, 5 x PFA-fitting with integrated ferrule, 2.3 mm OD, violet, 5 x PFA fitting with integrated ferrule, 1.6 mm OD, green, 10 x PFA fitting with integrated ferrule, 3.2 mm OD, blue, 10 x PFA blind plug, 2 x connector, straight, 6 to 8 mm ID, 1 x connector, curved, 5 to 11.5 mm ID, 1 x connector, angled, 9.5 to 10 mm ID 1 set
18626478 4 x Safety Cap III, GL45, 8 x air valve, 1 x Safety Waste Cap, GL45, 1 x Exhaust filter L, V3.0, with splash guard and change indicator, service life 12 months, 1 x 5 litre canister, PE-HD, 4 x Laboratory bottle DURAN, GL45, 1000 ml, clear glass and round, with cap and pouring ring , 5 x PFA-fitting with integrated ferrule, 2.3 mm OD, violet, 5 x PFA fitting with integrated ferrule, 1.6 mm OD, green, 10 x PFA fitting with integrated ferrule, 3.2 mm OD, blue, 10 x PFA blind plug, 2 x connector, straight, 6 - 8 mm ID, 1 x connector, curved, 5.0 - 11.5mm ID, 1 x connector, angled, 9.5 - 10 mm ID 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18636478 Supplier SCAT™ Supplier No. 307448
  • ONLINE EXCLUSIVE PRICE
    817.00€
    699.34€ / 1 feuilles
    Économisez 117.66€
    14% de réduction
Expédition estimée: 07-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
The all-round carefree package suitable for all HPLC and UHPLC systems.
  • Protects your system from contamination
  • Keeps solvent mixtures stable
  • High performance materials with excellent chemical resistance
The safety set connects tubing safely and blocks hazardous vapours. Air valves and exhaust filters provide safe pressure equalisation without under- or overpressure in solvent bottles and waste containers
Specifications
For Use With (Application) Liquid Chromatography
Description Electrostatic Conductive
For Use With (Equipment) Any HPLC System
Type Complete Solvent Collection System
Solvent Channels 4
Includes 4 x Safety Cap III, GL45, 8 x air valve, 1 x Safety Waste Cap, S60/61, electrostatic conductive, 1 x Exhaust filter L, V3.0, with splash guard and change indicator, service life 12 month, 1 x 5 litre canister, PE-HD-EL, 1 x Grounding cable, 2x clamp, L = 1,5 m, 4 x Laboratory bottle DURAN, GL45, 1000 ml, clear glass and round, with cap and pouring ring , 5 x PFA-fitting with integrated ferrule, 2.3 mm OD, violet, 5 x PFA fitting with integrated ferrule, 1.6 mm OD, green, 10 x PFA fitting with integrated ferrule, 3.2 mm OD, blue, 10 x PFA blind plug, 2 x connector, straight, 6 - 8 mm ID, 1 x connector, curved, 5.0 - 11.5mm ID, 1 x connector, angled, 9.5 - 10 mm ID