Learn More
Abcam Human BAZ1B (WSTF) knockout HeLa cell line
Product Code. 32142020 Shop All Abcam Products
Change view
Click to view available options
Quantity:
2 x 1,000,000 Cells
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32142020 2 x 1,000,000 Cells Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32142020 Supplier Abcam Supplier No. ab2649072x1000000Cells_
 Restricted Item
Expédition estimée: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
BAZ1B KO cell line available to order. KO validated by Western blot. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, Homozygous: 5 bp deletion in exon 2. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial. To order only knockout...
BAZ1B KO cell line available to order. KO validated by Western blot. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, Homozygous: 5 bp deletion in exon 2. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 2 x 1,000,000 Cells
Regulatory Status RUO