missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human beta-Glucuronidase/GUSB ELISA Kit
missing translation for 'orderingAttributeHoverText'
Quantity:
5 x 96 Tests
96 Tests
missing translation for 'unitSize'
Each
Pack of 96
Descrizione
Human beta-Glucuronidase/GUSB quantitates human beta-Glucuronidase/GUSB in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human beta-Glucuronidase/GUSB.
The GUSB gene encodes beta-glucuronidase (EC 3.2.1.31), a lysosomal hydrolase involved in the stepwise degradation of glucuronic acid-containing glycosaminoglycans. It is a tetrameric glycoprotein composed of identical subunits. The GUSB gene is mutated in mucopolysaccharidosis type VII (MPS7; MIM 253220).
Specifica
Specifica
| Accession Number | Ac2-223; adipose storage deficiency; AI747421; asd; beta glucuronidase; beta-D-glucuronidase; beta-G1; Beta-glucuronidase; BG; g; glucuronidase beta; glucuronidase, beta; Gur; Gus; GUSB; Gus-r; Gus-s; Gus-t; Gus-u; Gut; MPS7 |
| Assay Range | 0.18 to 40 ng/mL |
| Assay Sensitivity | 0.18 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 2990 |
| Gene Symbol | GUSB |
| Interassay CV | <12% |
| Vedi altri risultati |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto