Learn More
Gibco™ Human Dendritic Cell Cytokine Package, PeproTech®
Product Code. 17813703
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17813703 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17813703 Supplier Gibco™ Supplier No. HDC
 Restricted Item
Expédition estimée: 14-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
The Human Dendritic Cell Cytokine Package contains the key components required for the culturing of human dendritic cells.
IL-4 is a pleiotropic cytokine that regulates diverse T and B cell responses including cell proliferation, survival, and gene expression. Produced by mast cells, T cells, and bone marrow stromal cells, IL-4 regulates the differentiation of naive CD4+ T cells into helper Th2 cells.
GM-CSF is a hematopoietic growth factor that stimulates the development of neutrophils and macrophages and promotes the proliferation and development of early erythroid megakaryocytic and eosinophilic progenitor cells.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Functional Assay
Product Type Dendritic Cell Cytokine Package
Quantity 1 Kit
Content And Storage 100 μg Human IL-4 (Cat. No. 200-04-100 UG), 100 μg Human GM-CSF (Cat. No. 300-03-100 UG)
Species Human