Learn More
Cytiva HyClone™ CDM4PERMAb cell culture media
Product Code. 16407675
Change view
Click to view available options
Quantity:
1000 mL
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16407675 1000 mL 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16407675 Supplier Cytiva Supplier No. SH30871.02
  • ONLINE EXCLUSIVE PRICE
    116.00€
    102.00€ / 1L
    Économisez 14.00€
    12% de réduction
 Restricted Item
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Chemically defined Animal-Derived Component-Free (ADCF) media for use with PER.C6 cells
  • Chemically defined and animal-derived component-free (ADCF) formulation
  • This high-performance cell culture medium has been developed through the Metabolic Pathway Design process to increase process yields in the production of human antibodies and recombinant proteins using PER.C6 technology
  • The medium has been successfully tested in a variety of applications, including fed-batch bioreactor cultures
  • Designed for high cell yield and recombinant protein production
  • Allows for direct or sequential adaptation
  • Component traceability
  • Manufactured according to cGMP guidelines
  • TRUSTED_SUSTAINABILITY
    Specifications
    Classification None
    Form Liquid
    Product Type Protein Production Medium
    Sterility Sterile
    Sterilization Method 0.2 μm Sterile-filtered
    With Additives None
    Without Additives No Glutamine, No Phenol Red, No HEPES
    Quantity 1000 mL
    Format Bottle
    Content And Storage 2° to 8°C, away from light