Learn More
Neogen Hydrated-Sponge
Product Code. 17298510
Encompass
Change view
Click to view available options
Includes:
Sponge, Gloves
Saturation Solution:
10 mL Neutralizing Buffer
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Saturation Solution Unit Size
17298510 Sponge, Gloves 10 mL Neutralizing Buffer 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17298510 Supplier Neogen Supplier No. 7000002888
 Restricted Item
Expédition estimée: 02-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Uses simplified packaging to minimize handling and preparation, and reduces waste.
  • Ready-to-use, pre-hydrated sponges for easy and convenient environmental and food sample collection.
  • Durable sponge withstands scrubbing.
  • Sponge bag with write-on label for ease in labeling samples.
  • Includes two decontaminated gloves for safety and convenience.
Specifications
Product Type Hydrated Sponge
Length (English) 3 in.
Width (English) 1.5 in.
Packaging Quantity 100/Cs.
Saturation Solution 10 mL Neutralizing Buffer
Includes Sponge, Gloves