Description
Sophisticated features ensure superior wearer comfort even during long periods of use.
General features
- Safety spectacles with soft, flexible nose piece
- Incl. additional frame for even greater wearer comfort and a secure fit
- The uvex supravision excellence lens coating provides permanent anti-fog protection on the inside and is chemical- and scratch-resistant on the outside
- Metal-free
- Frame color: anthracite, blue
- PC lens: clear
Protective features
- Certified in line with EN 166 (personal eye protection) and EN 170 (UV filters)
- Marking: W 166 FT CE – 2C-1,2 W 1 FTKN CE
- Reliable UV400 protection
Comfort features
- Soft, flexible nose pad for maximum wearer comfort and a non-slip fit
- X-tended eyeshield provides protection against dust, dirt and flying particles
- X-tended sideshield provides further protection without restricting peripheral vision
- 3-position inclination allows wearers to achieve a comfortable fit
- Extendable x-twist side arm technology offers flexible, convenient adjustment to any head shape
- Soft ear-pieces for a pressure-free, non-slip fit
Specifications
Specifications
| Lens Type | Chemical-resistant |
| Lens Coating | Anti-fog, Scratch-resistant |
| Color | Anthracite, Blue |
| Certifications/Compliance (Europe) | EN 166, EN 170 |
| Lens Tint | Clear |
| Product Type | Ergonomic Safety Glasses |
| Material | Polycarbonate |
| Disposable | Reusable |
| Lens Material | Polycarbonate |
| Frame Material | Plastic |
| Show More |
Product Title
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?