Learn More
Invitrogen™ eBioscience™ IHC Antigen Retrieval Solution - Low pH (10X)
Product Code. 15566276
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15566276 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15566276 Supplier Invitrogen™ Supplier No. 00495558
 Restricted Item
En stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy
Designed for use during the heat induced epitope retrieval (HIER) step prior to immunohistochemistry on formalin-fixed paraffin-embedded tissue sections. Used in combination with heat has been shown to restore the antigenicity of proteins modified during the formalin fixation of tissue.
  • Supplied as a 10x stock solution
  • Dilution of this buffer to 1x using reagent-grade water results in a 10 mM Sodium Citrate solution, pH 6.0
TRUSTED_SUSTAINABILITY
Specifications
Product Type IHC Antigen Retrieval Solution
Quantity 500 mL
For Use With (Application) Immunohistochemistry (IHC)
Product Line eBioscience
Form Liquid
For Research Use Only. Not for use in diagnostic procedures.