Learn More
Proteintech IHCeasy GLUL Ready-To-Use IHC Kit
Product Code. 17456743 Shop All Proteintech Products
Change view
Click to view available options
Packaging Quantity:
50 Tests
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Quantity Unit Size
17456743 50 Tests 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17456743 Supplier Proteintech Supplier No. KHC003750T
 Restricted Item
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
A ready-to-use IHC kit for the staining of Glutamine Synthetase (GS or GLUL).
  • This kit provides all reagents, from antigen retrieval to cover slip mounting, that require little to no diluting or handling prior to use.
  • Simply apply the reagents to your sample slide according to the protocol and you're steps away from obtaining high-quality IHC data.
Specifications
Target Species Human, Rat, Mouse
Sample Type FFPE Tissue
Content And Storage Store at 2°C to 8°C
Packaging Quantity 50 Tests
Sufficient For 50 Reactions