Learn More
Hirschmann™ Imhoff Sedimentation Cone
Product Code. 13158139
Change view
Click to view available options
Volume (Metric):
1000 mL
Unit Size:
Pack of 3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
13158139 1000 mL Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13158139 Supplier Hirschmann™ Supplier No. 7720193
  • ONLINE EXCLUSIVE PRICE
    160.25€
    148.68€ / 3 pièces
    Économisez 11.57€
    7% de réduction
View all promotions
Expédition estimée: 21-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Unbreakable, transparent Sedimentation Cone acc. to Imhoff
  • SAN
  • Imhoff type
  • Transparent
  • Unbreakable
  • Moulded graduation
  • Screw cap at typ for emptying
Specifications
Description Sedimentation Cone, acc. to Imhoff
Volume (Metric) 1000 mL
Graduated Raised scale
Material SAN
No. per Pack 3