Learn More
Imidapril Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Imidapril Hydrochloride |
| CAS | 89396-94-1 |
| Molecular Formula | C20 H27 N3 O6 . Cl H |
| Quantity | 100 mg |
| Synonym | (4S)-3-[(2S)-2-[[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-1-oxopropyl]-1-methyl-2-oxo-4-imidazolidinecarboxylic Acid Hydrochloride |
| InChI Formula | InChI=1S/C20H27N3O6.ClH/c1-4-29-19(27)15(11-10-14-8-6-5-7-9-14)21-13(2)17(24)23-16(18(25)26)12-22(3)20(23)28;/h5-9,13,15-16,21H,4,10-12H2,1-3H3,(H,25,26);1H/t13-,15-,16-;/m0./s1 |
| SMILES | Cl.CCOC(=O)[C@H](CCc1ccccc1)N[C@@H](C)C(=O)N2[C@@H](CN(C)C2=O)C(=O)O |
| IUPAC Name | (4S)-3-[(2S)-2-[[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino]propanoyl]-1-methyl-2-oxoimidazolidine-4-carboxylic acid;hydrochloride |
| Molecular Weight (g/mol) | 441.91 |
| Formula Weight | 441.1667 |