Learn More
Imidazo[1,2-a]pyridin-2-ylmethanol, 90%, BLD Pharm™
Product Code. 31450871
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31450871 250 mg
31441310 100 mg
31434395 1 g
31447575 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31450871 Supplier BLD Pharm Supplier No. BD134383250mg
 Restricted Item
Expédition estimée: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 82090-52-6
Molecular Formula C8H8N2O
MDL Number MFCD06797471
Quantity 250 mg
SMILES OCC1=CN2C=CC=CC2=N1
Molecular Weight (g/mol) 148.16
Percent Purity 90%
Chemical Name or Material Imidazo[1,2-a]pyridin-2-ylmethanol