Learn More
Iminodibenzyl (10,11-Dihydro-5H-dibenzo[b,f]azepine), LoGiCal
Product Code. 30460264
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30460264 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30460264 Supplier LoGiCal Supplier No. LGCFOR0076.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Especificaciones
Chemical Name or Material Iminodibenzyl (10,11-Dihydro-5H-dibenzo[b,f]azepine)
CAS 494-19-9
Analyte or Component Names Iminodibenzyl (10,11-Dihydro-5H-dibenzo[b,f]azepine)
Shipping Condition Room Temperature
Molecular Formula C14 H13 N
Certifications/Compliance ISO 9001
Synonym Trimipramine Maleate Imp. B (EP), Trimipramine Imp. B (EP), Iminodibenzyl, 10,11-Dihydro-5H-dibenzo[b,f]azepine, Clomipramine Hydrochloride Imp. E (EP), Trimipramine Maleate Imp. F (EP), Carbamazepine Imp. E (EP), Carbamazepine Impurity E, Clomipramine Hydrochloride Impurity E, Trimipramine Maleate Impurity F, Clomipramine Impurity E, Trimipramine Impurity F
InChI Formula InChI=1S/C14H13N/c1-3-7-13-11(5-1)9-10-12-6-2-4-8-14(12)15-13/h1-8,15H,9-10H2
Solution Type Neat
SMILES C1Cc2ccccc2Nc3ccccc13
IUPAC Name 6,11-dihydro-5H-benzo[b][1]benzazepine
Industry Type Clinical Testing
Molecular Weight (g/mol) 195.26
Quantity 10 mg
Formula Weight 195.1048 g/mol
Mostrar más Mostrar menos
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.