Learn More
Motic™ Immersion Oil
Product Code. 15485092
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15485092 5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15485092 Supplier Motic™ Supplier No. 1101001300011
 Restricted Item
En stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy
Transparent oil of high refractive index for use with oil immersion objectives.
  • High refractive index.
  • Transparent.
  • Increases the numerical aperture of the objective lens.
  • Increases resolving power of a microscope.
Specifications
For Use With (Application) Oil immersion objectives
Quantity 5 mL
compliance-icons compliance-icons
Product Identifier
  • cargille immersion oil
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P391-Collect spillage.
Supplemental information
  • MIXTURE LIST-Contain: paraffins (petroleum), hydrogenated terphenyls, butene, terphenyl