missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B Braun™ Inerocan™ Safety Winged IV Catheter
Description
- Promotes first-stick success and clinician confidence
- Easier to introduce into the vein compared to the leading IV catheter
- Features a universal bevel that is designed for a wider choice of insertion angles and produces less tearing
- Winged hub designed to improve stabilization and lead to longer indwell performance, less labor and materials spent on restarts and greater convenience
- Passive safety design ensures that the safety mechanism is always activated and prevents re-insertion of the stylet
- Lighter, more compact design can potentially lower shipping, storage and waste disposal costs
- PVC- and latex-free
- Manufactured from PUR (polyurethane)
- Supplied in counts of 50
Specifications
Specifications
| Product Type | Safety IV Catheter |
| Color | Pink |
| Material | Polyurethane |
| Needle Gauge | 20 G |
| Sterility | Sterile |
| For Use With (Application) | Reducing needlestick injuries |
| Includes | Winged hub |
| Length (Metric) Needle | 32 mm |
| Quantity | 50/Pk. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?