Learn More
Abeba™ Insoles 3553
Product Code. 15690128
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15690128 42 1 pair
15630128 36 1 pair
15640128 37 1 pair
15650128 38 1 pair
15660128 39 1 pair
15670128 40 1 pair
15680128 41 1 pair
15600138 43 1 pair
15610138 44 1 pair
15620138 45 1 pair
15630138 46 1 pair
15640138 47 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Product Code. 15690128 Supplier Abeba™ Supplier No. 3553/42
  • ONLINE EXCLUSIVE PRICE
    21.27€
    14.79€ / 1 paires
    Économisez 6.48€
    30% de réduction
Expédition estimée: 17-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
ESD dissipative insoles include antibacterial and antifungal properties due to active carbon.
  • Breathable textile
  • Blue
  • ESD dissipative according to DIN EN 61340
  • Antistatic
  • Areal pressure absorption, climatising, comfortable
  • Benefit for joints, musculature, ligaments and spine
Specifications
Color Blue
Material Textile
Size 42
Style 3553