Learn More
Invitrogen™ pcDNA™3.1/myc-His A, B, & C Mammalian Expression Vectors
Product Code. 10778994
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10778994 20 μg 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10778994 Supplier Invitrogen™ Supplier No. V80020
  • ONLINE EXCLUSIVE PRICE
    1111.80€
    958.50€ / 1 kits
    Économisez 153.30€
    13% de réduction
 Restricted Item
Expédition estimée: 09-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Contain a strong promoter and a selection marker for generating stable cell lines
  • Includes an epitope tag for easy detection with monoclonal antibody and rapid purification on nickel-chelating resin
  • Each vector is available in three reading frames to simplify cloning in-frame with fusion tag
Constitutive Expression, Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis
TRUSTED_SUSTAINABILITY
Specifications
Constitutive or Inducible System Constitutive
Delivery Type Transfection
Promoter CMV
Product Type Mammalian Expression Vector
Selection Agent (Eukaryotic) Geneticin™ (G-418)
Content And Storage Vectors (and appropriate expression control) are supplied supercoiled and lyophilized. Stored at -20°C. All vectors are guaranteed stable for 6 months when properly stored.
Protein Tag His Tag (6x), c-Myc Epitope Tag
Cloning Method Restriction Enzyme/MCS
Quantity 20 μg
Vector pcDNA
Product Line pcDNA
For Use With (Application) Constitutive Expression
Show More Show Less
For Research Use Only. Not for use in diagnostic procedures.