Learn More
Invitrogen™ Yeast Deletion Matα, Complete Set
Product Code. 10682564
Change view
Click to view available options
Quantity:
53 Plates
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10682564 53 Plates Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10682564 Supplier Invitrogen™ Supplier No. 95401.H3
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Yeast deletion library created by a collaboration of eight North American and eight European laboratories composing the Saccharomyces Genome Deletion Project
Each yeast deletion clone is supplied as a stab culture in yeast peptone dextrose (YPD) agar containing Geneticin™ (G418).
Order Info
Shipping Condition: Dry Ice
TRUSTED_SUSTAINABILITY
Specifications
Product Type Yeast Deletion Mat
Libraries Yeast deletion library created by a collaboration of 8 North American and 8 European laboratories composing the Saccharomyces Genome Deletion Project
Culture Type Stab culture in yeast peptone dextrose (YPD) agar containing Geneticin™ (G418)
Format Complete Set
Quantity 53 Plates