Learn More
Fisherbrand™ Ion Selective Electrode Iodide
Artikelnummer. 11709718 Shop alle Fisherbrand Traceable produkter
Skift visning
Klik for at se tilgængelige muligheder
pH Range:
0 to 14
Pakningsstørrelse:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. pH Range Pakningsstørrelse
11709718 0 to 14 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 11709718
  • ONLINE EXCLUSIVE PRICE
    936.00€
    308.21€ / 1 pièces
    Économisez 627.79€
    67% de réduction
Se alle kampagner
 Restricted Item
Expédition estimée: 20-01-2027
Læg i indkøbskurv
Læg i indkøbskurv
Combined ion-sensitive iodide electrode with solid membrane interchangeable flat surface is ideally delivered with membrane, electrolytic solution and gasket.
  • Combined ion-sensitive electrode with solid membrane interchangeable flat surface
  • The ion to be detected passes through the membrane generating a potential difference proportional to the ionic concentration
  • Analysis time typically less than one minute
  • Sample preparation steps such as filtering or distillation rarely necessary
  • Calibration should be carried out as often as possible
  • (less once a day) at the same temperature as the sample
  • Measuring temperature 0 to 50 ° C
Tekniske data
Description Ion Selective Electrode Iodide
pH Range 0 to 14
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.