Learn More
Ambion™ iPrep™ Card: Total RNA
Product Code. 10419793
Change view
Click to view available options
Quantity:
1 Card
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10419793 1 Card Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10419793 Supplier Ambion™ Supplier No. IS10014
 Restricted Item
Expédition estimée: 07-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
iPrep™ Card: Total RNA
The iPrep™ Protocol Card is a flash, 512 KB memory card. Each iPrep™ Protocol card is pre-programmed with the purification protocol that directs the volume of reagents used and incubation time.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at room temperature.
For Use With (Equipment) iPrep™
Product Line iPrep
Quantity 1 Card
Shipping Condition Room Temperature
For Research Use Only. Not for use in diagnostic procedures.