Learn More
Thermo Scientific™ iSC-65 Autosampler
Product Code. 17854185 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
iCAP PRO X/XP/XPS ICP-OES, iCAP MX and iCAP Qnova Series ICP-MS
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17854185 iCAP PRO X/XP/XPS ICP-OES, iCAP MX and iCAP Qnova Series ICP-MS Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 17854185 Supplier Thermo Scientific™ Supplier No. BRE0030065
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Accelerate elemental sample throughput by automating your trace elemental analysis workflow with a Thermo Scientific™ iSC-65 Autosampler.
Achieve robust, worry-free automation of your high-throughput trace elemental analysis workflow with the Thermo Scientific™ iSC-65 Autosampler. The iSC-65 Autosampler is built to thrive in high-throughput laboratories and difficult environments without downtime or contributing contamination to your time-critical trace elemental samples. The quiet, smooth operation will blend into the background of your working environment. Acid and solvent-resistant construction materials assure robust, reliable, precise operation.
View product tour
Overview
  • Compatible with Thermo Scientific trace elemental analysis instruments: Thermo Scientific™ iCAP™ MX and iCAP™ Qnova Series ICP-MS (iCAP MSX, MTX, Q, RQ, RQplus, TQ, TQe, and TQs) and Thermo Scientific™ iCAP™ PRO Series ICP-OES (iCAP PRO X, PRO XP, and PRO XPS)
  • Four rack capacity, each supporting 21, 24, 40, 60, or 90 vial positions
  • Ten-position 50 mL round bottom or flat bottom standard rack
  • Variable speed two-channel rinse pump; luer lock rinse station fittings
  • X, Y, and Z drive obstruction detection
  • Step Ahead – programmable rinse sequence begins during sample analysis, saving time
  • Multi-color LED display
  • Complete integration with Thermo Scientific™ Qtegra™ ISDS Software, including alignment and all analysis parameters
  • Ability to set depth individually for each vial location
  • Ethernet communications
  • Two auxiliary ports
iSC-65 Autosampler attained an ACT label
Know your lab's environmental impact with the iSC-65 Autosampler, recipient of the ACT label from My Green Lab. The ACT Environmental Impact Factor Label is the world's premier eco-label for laboratory products. The program ensures Accountability, Consistency, and Transparency in the reporting of environmental impact data to enable sustainable laboratory procurement. It was designed by both scientists and procurement specialists to provide clear, third-party verified information about the sustainability profile of laboratory products. By providing needed transparency around manufacturing, energy and water use, packaging, and end-of-life impacts, ACT makes it easy to choose environmentally preferable products and reduce the carbon impact of laboratory supply chains.
Specifications
For Use With (Equipment) iCAP PRO X/XP/XPS ICP-OES, iCAP MX and iCAP Qnova Series ICP-MS
Description iSC-65 Autosampler
Voltage 100/240 V