Learn More
STEMCELL Technologies Iscove's Modified Dulbecco's Medium
Product Code. 17108091
Encompass
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17108091 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17108091 Supplier STEMCELL Technologies Supplier No. 36150
 Restricted Item
Expédition estimée: 20-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Iscove's Modified Dulbecco's Medium (IMDM) with 25 mM HEPES
  • Iscove's Modified Dulbecco's Medium (IMDM) with 25 mM HEPES is recommended for a wide variety of cell culture applications
  • Selection of suitable nutrient medium is dependent on the type of cell, conditions of culture, and degree of chemical definition required for the cell culture application
Specifications
For Use With (Application) Cell Culture
Quantity 500 mL