Learn More
Isepamicin Sulfate, TRC
Product Code. 30011242
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011242 250 mg 250mg
30003909 2.5 g 2.5g
30005278 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011242 Supplier Toronto Research Chemicals Supplier No. TRCI927013250MG
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 67814-76-0
Molecular Formula C22H43N5O12 • H2O4S
Molecular Weight (g/mol) 569.609808
Synonym D-Streptamine, O-6-amino-6-deoxy-α-D-glucopyranosyl-(1→4)-O-[3-deoxy-4-C-methyl-3-(methylamino)-β-L-arabinopyranosyl-(1→6)]-N1-(3-amino-2-hydroxy-1-oxopropyl)-2-deoxy-, (S)-, sulfate (salt), D-Streptamine, O-6-amino-6-deoxy-α-D-glucopyranosyl-(1→4)-O-[3-deoxy-4-C-methyl-3-(methylamino)-β-L-arabinopyranosyl-(1→6)]-N1-[(2S)-3-amino-2-hydroxy-1-oxopropyl]-2-deoxy-, sulfate (salt) (9CI), Exacin, Isepacin
SMILES CN[C@@H]1[C@@H](O)[C@H](OC[C@]1(C)O)O[C@@H]2[C@@H](O)[C@H](O[C@H]3O[C@H](CN)[C@@H](O)[C@H](O)[C@H]3O)[C@@H](N)C[C@H]2NC(=O)[C@@H](O)CN.O[S](O)(=O)=O
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C22H43N5O12.H2O4S/c1-22(35)6-36-20(15(33)18(22)26-2)39-17-8(27-19(34)9(28)4-23)3-7(25)16(14(17)32)38-21-13(31)12(30)11(29)10(5-24)37-21;1-5(2,3)4/h7-18,20-21,26,28-33,35H,3-6,23-25H2,1-2H3,(H,27,34);(H2,1,2,3,4)/t7-,8+,9-,10+,11+,12-,13+,14-,15+,16+,17-,18+,20+,21+,22-;/m0./s1
Chemical Name or Material Isepamicin Sulfate