Learn More
Brady™ ISO 7010 Sign - No open flame, Fire, open ignition source and smoking prohibited
Product Code. 15604587 Shop All Brady Products
Change view
Click to view available options
Material:
Polypropylene
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
15604587 Polypropylene Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15604587 Supplier Brady™ Supplier No. 822150
  • ONLINE EXCLUSIVE PRICE
    14.50€
    12.26€ / 1 pièces
    Économisez 2.24€
    15% de réduction
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Prevent costly accidents with ISO 7010 safety signs for optimal impact and recognizability.
  • Polypropylene
  • For indoor and outdoor use
Specifications
Certifications/Compliance ISO 7010
Alternate Part Number PIC P003-Dia 200-PP-CRD/1
Material Polypropylene
Quantity 1 Ea.
For Use With (Application) Indoor and Outdoor
Thickness 1.501 mm
Temperature Range (Metric) -40°C to 120°C
Shape Round
Languages Global
Color Black