Learn More
Brady™ ISO Horizontal Safety Sign - No Smoking
Product Code. 15707659 Shop All Brady Products
Change view
Click to view available options
Dimensions (L x W):
105 x 297 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
15707659 105 x 297 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15707659 Supplier Brady™ Supplier No. 139089
  • ONLINE EXCLUSIVE PRICE
    8.45€
    7.12€ / 1 pièces
    Économisez 1.33€
    15% de réduction
Expédition estimée: 14-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Safety sign for indicating mandatory action.
  • Sign compliant to ISO 7010
  • Improves workplace safety
Specifications
Alternate Part Number P/P002/FR76-SA-297x105/1-B
Finish Gloss
Material Laminated Polyester
Adhesive Type Permanent Acrylic
Shape Rectangular
Languages French
Dimensions (L x W) 105 x 297 mm
Displayed Text Défense de fumer
Color Red on White