Learn More
Brady™ ISO Safety Sign without Text - Danger: Pressurized Cylinder
Product Code. 15881537 Shop All Brady Products
Change view
Click to view available options
Material:
Laminated Polyester
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
15881537 Laminated Polyester Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15881537 Supplier Brady™ Supplier No. 830611
  • ONLINE EXCLUSIVE PRICE
    10.00€
    8.45€ / 1 pièces
    Économisez 1.55€
    15% de réduction
 Restricted Item
Expédition estimée: 08-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Safety sign for hazard identification.
  • Sign compliant to ISO 7010
  • Improves workplace safety
Specifications
Alternate Part Number PIC W029-Tri 100-PE-SHEET/3
Finish Gloss
Material Laminated Polyester
Adhesive Type Permanent Acrylic
Shape Triangular
Dimensions (L x W) 87 x 100 mm
Color Black on Yellow