missing translation for 'onlineSavingsMsg'
Learn More

(±)-Isoborneol, 95%

Product Code. 11419067
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419067 100 g 100g
11409067 25 g 25g
11488910 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11419067 Supplier Thermo Scientific Alfa Aesar Supplier No. B20632.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 124-76-5 | C10H18O | 154.253 g/mol

Used as a synthetic flavor, as a moth repellent, cold sore topical medication, muscle liniment, and steam-inhaled cough suppressant. It is also used in Used in perfumes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a synthetic flavor, as a moth repellent, cold sore topical medication, muscle liniment, and steam-inhaled cough suppressant. It is also used in Used in perfumes.

Solubility
Insoluble in water. Soluble in dipropylene glycol, ethyl alcohol (1gm. in 2ml. 70% alcohol).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-76-5
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
MDL Number MFCD00074821
InChI Key DTGKSKDOIYIVQL-SZBHIRRCSA-N
Synonym dl-isoborneol
PubChem CID 126961757
IUPAC Name (1R,3R)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-ol
SMILES CC1(C2CCC1(C(C2)O)C)C

Specifications

Melting Point ∼210°C (sublimation)
Flash Point 74°C (165°F)
Odor Strong
Quantity 100 g
UN Number UN1325
Beilstein 4126091
Merck Index 14,5128
Formula Weight 154.25
Percent Purity 95%
Chemical Name or Material (±)-Isoborneol
compliance-icons compliance-icons
Product Identifier
  • Isoborneol
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H228-Flammable solid.
  • H315-Causes skin irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Isoborneol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.