missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Isomaltotriose, 97%

Product Code. 18988390
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
18988390 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18988390 Supplier Thermo Scientific Chemicals Supplier No. J64186.ME
 Restricted Item
Expédition estimée: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3371-50-4 | C18H32O16 | 504.438 g/mol

Isomaltotriose is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isomaltotriose is used as pharmaceutical intermediate.

Solubility
Soluble in water

Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3371-50-4
Molecular Formula C18H32O16
Molecular Weight (g/mol) 504.438
MDL Number MFCD00070623
InChI Key FBJQEBRMDXPWNX-IGSOQDMUSA-N
Synonym isomaltotriose, 3r,4s,5s,6r-6-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy methyl oxan-2-yl oxy methyl oxane-2,3,4,5-tetrol, isomaltotriose, dp3, glc a1-6 glc a1-6 glc, isomaltotriose tlc, isomaltotriose, dp3, analytical standard, alpha-d-glucosyl-1->6-alpha-d-glucosyl-1->6-d-glucose, 6-o-6-o-alpha-d-glucopyranosyl-alpha-d-glucopyranosyl-d-glucopyranose, d-glucose, o-.alpha.-d-glucopyranosyl-1->6-o-.alpha.-d-glucopyranosyl-1->6, 3r,4s,5s,6r-6-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-2-yl oxymethyl oxane-2,3,4,5-tetrol
PubChem CID 133126476
IUPAC Name (3S,4R,5R,6S)-6-[[(2R,3S,4R,5R,6S)-3,4,5-trihydroxy-6-[[(2R,3S,4R,5R,6S)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxymethyl]oxan-2-yl]oxymethyl]oxane-2,3,4,5-tetrol
SMILES C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OCC3C(C(C(C(O3)O)O)O)O)O)O)O)O)O)O)O

Specifications

Color White
Quantity 500 mg
Solubility Information Soluble in water
Formula Weight 504.44
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Isomaltotriose

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.