Learn More
Isopropylamine Hydrobromide, 98%, BLD Pharm™
Product Code. 30968315
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30968315 1 g 1g
30966306 5 g 5g
30957939 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30968315 Supplier BLD Pharm Supplier No. BD008050751g
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 29552-58-7
Molecular Formula C3H10BrN
MDL Number MFCD22419143
Quantity 1 g
SMILES NC(C)C.[H]Br
Molecular Weight (g/mol) 140.02
Percent Purity 98%
Chemical Name or Material Isopropylamine Hydrobromide