Learn More
Itraconazole, 98%
Description
Itraconazole is a triazole antifungal agent. It is used to inhibit cytochrome P-450-dependent enzymes and ergosterol synthesis. It has been used against histoplasmosis, blastomycosis, cryptococcal meningitis, and aspergillosis. It?s different formulations are used to study Candida strains in murine invasive infections. It has been used to study proliferative changes of the forestomach mucosa in alloxan-induced diabetic rats..
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Itraconazole is a triazole antifungal agent. It is used to inhibit cytochrome P-450-dependent enzymes and ergosterol synthesis. It has been used against histoplasmosis, blastomycosis, cryptococcal meningitis, and aspergillosis. It′s different formulations are used to study Candida strains in murine invasive infections. It has been used to study proliferative changes of the forestomach mucosa in alloxan-induced diabetic rats..
Solubility
Insoluble in water. Solube in chloroform at 50 mg/ml. Slightly soluble in ethanol or methanol
Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Specifications
Specifications
| Type | Itraconazole |
| Physical Form | Powder |
| Melting Point | 166°C |
| Color | White |
| Quantity | 1 g |
| Flash Point | >110°C (230°F) |
| Merck Index | 14,5245 |
| Solubility Information | Insoluble in water; Solube in chloroform at 50mg/ml; Slightly soluble in ethanol or methanol |
| Formula Weight | 705.63 |
| Percent Purity | 98% |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.