Learn More
GPE Scientific™ J Young NMR Valve with Tube Attached
Product Code. 15854874
Change view
Click to view available options
Type:
NMR Valve with Tube, 200 MHz
NMR Valve with Tube, 500 MHz
Length (English):
7 in.
8 in.
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Length (English) Unit Size
15854874 NMR Valve with Tube, 200 MHz 7 in. Each
15864874 NMR Valve with Tube, 200 MHz 8 in. Each
15874874 NMR Valve with Tube, 500 MHz 7 in. Each
15884874 NMR Valve with Tube, 500 MHz 8 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15854874 Supplier GPE Scientific™ Supplier No. S5200JY7YELLOW
  • ONLINE EXCLUSIVE PRICE
    101.01€
    93.23€ / 1 pièces
    Économisez 7.78€
    7% de réduction
Expédition estimée: 11-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
GPE Scientific™ J Young NMR Valve with Tube Attached allows you to handle your NMR sample without flame sealing your tubes.
  • Handle your NMR sample without flame sealing your tubes.
  • Teflon covered O-ring eliminates material incompatibilities.
  • Completely greaseless Teflon assembly means the tube is easy to use and to disassemble for cleaning.
  • Includes female joint for quick attachment to your vacuum rack.
  • Yellow piston
  • Negative pressure/high vacuum.
  • Leak tested to 10-9mbar liters/sec.
  • Internal positive pressure 5bar, 72psi
Specifications
Type NMR Valve with Tube, 200 MHz
Includes 5mm NMR Valve
Length (English) 7 in.