Learn More
GPE Scientific™ J Young Schlenk Flask, Round Bottom with Fine Thread Tap and Socket
Product Code. 15884804
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15884804 500 mL Each
15854804 25 mL Each
15874804 50 mL Each
15834804 100 mL Each
15864804 250 mL Each
15844804 1000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15884804 Supplier GPE Scientific™ Supplier No. JRF/500
Expédition estimée: 09-02-2027
Add to basket
Add to basket
This item is not returnable. View return policy
GPE Scientific™ J Young Schlenk Flask, Round Bottom with Fine Thread Tap and Socket is specially designed for use with Schlenk lines and vacuum manifolds.
  • Solidly constructed for use with vacuum manifolds
  • Designed for use with Schlenk lines
  • Includes fine thread tap and socket
Specifications
Capacity (Metric) 500 mL
For Use With (Equipment) Schlenk lines and vacuum manifolds
Includes Fine thread tap and socket
Socket Joint Size: 24
Bottom Shape Round