Learn More
Justrite™ Cease-Fire™ Head
Product Code. 11973669
Change view
Click to view available options
Opening Size (English):
5.25 in.
7.8 in.
Diameter (English) Outer:
15.12 in.
24 in.
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Opening Size (English) Diameter (English) Outer Unit Size
11973669 7.8 in. 24 in. Each
17849343 5.25 in. 15.12 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11973669 Supplier Justrite™ Supplier No. 26555
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
Convert standard metal drum into fire-safe waste receptacle
  • Install easily on existing metal drums
  • Aluminum construction is virtually corrosion free for easy clean-up
Certifications
FM approved
Specifications
Certifications/Compliance FM, UL
Diameter (Metric) Outer 61 cm
Diameter (English) Outer 24 in.
Opening Size (English) 7.8 in.
Opening Size (Metric) 20 cm
Quantity 1 Ea.
For Use With (Application) Install easily on existing metal drums
For Use With (Equipment) Cease-fire container, 55 gal. container size
Material Aluminum
Product Type Cease-Fire Head