Learn More
behr Labor-Technik™ K 20 Model Standard Kjeldahl Block Digestion System
Product Code. 10186394
Change view
Click to view available options
Capacity (Metric):
250 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10186394 250 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 10186394 Supplier behr Labor-Technik™ Supplier No. B00632831
  • ONLINE EXCLUSIVE PRICE
    8336.55€
    6798.53€ / 1 pièces
    Économisez 1538.02€
    18% de réduction
Expédition estimée: 04-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Complete system with digestion vessel, frame and extraction hood. With 8, 12 or 20 sample positions for standard Kjeldahl digestion vessels with a volume of 250 mL. The block casing as well as the rack for the digestion devices and the hood are made of acid-resistant, non-corrosive stainless steel.
  • High-performance heating and extraction hood with exhaust collector
    • The progress of the digestion can be observed via the removable viewing door on the insert rack
      • behr one-button operation for extremely easy and quick programming
        • Menu navigation in country language
          • 25 freely configurable programs for block temperature and digestion time
            • Storeable applications
              • USB interface
              • Complete system with digestion vessel, frame and extraction hood.
              • Stable and robust design.
Specifications
Product Type Kjeldahl Block Digestion System
Wattage 1000 W
Capacity (Metric) 250 mL
No. of Places 20
Temperature (Metric) Maximum 430°C