Learn More
Lonza™ KGM™ Keratinocyte Growth Medium SingleQuots™ Supplements and Growth Factors
Product Code. 11675490
Change view
Click to view available options
Cell Type:
Keratinocyte Cells
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cell Type Unit Size
11675490 Keratinocyte Cells Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11675490 Supplier Lonza™ Supplier No. CC4131
  • ONLINE EXCLUSIVE PRICE
    255.00€
    201.85€ / 1 pièces
    Économisez 53.15€
    20% de réduction
 Restricted Item
Expédition estimée: 17-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Supplements and growth factors kit for use with keratinocyte basal medium.
  • Separately sold SingleQuot™ kit for supplementing basal medium.
  • Part of the full KGM™ Keratinocyte Cell Growth Medium BulletKit™ (catalog number CC-3111), which consists of basal medium and supplements packaged together.
  • Includes the following:
    • BPE Low Protein - non specific
    • HEGF
    • Recombinant Human Insulin 0.5%
    • Hydrocortisone Solution
    • GA-1000
TRUSTED_SUSTAINABILITY
Specifications
Cell Type Keratinocyte Cells
Product Type Supplements and growth factors
Content And Storage BPE Low Protein - non specific, HEGF, Recombinant Human Insulin 0.5%, Hydrocortisone Solution, GA-1000