Learn More
DiscoverX KILR™ Retroparticles for Adherent Cells (Hygromycin B)
Product Code. 17304839
Encompass
Change view
Click to view available options
Quantity:
4 Vials
Unit Size:
4 ampoules
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17304839 4 Vials 4 ampoules
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17304839 Supplier DiscoverX Supplier No. 970005
 Restricted Item
Expédition estimée: 07-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
The KILR™ Retroparticles for Adherent Cells (Hygromycin B) are retroparticles for transducing cells of interest with PL to allow for cytotoxicity assays. Contains 4 vials of cryopreserved retroparticles. Resists Hygromycin B.
The KILR™ Retroparticles for Adherent Cells (Hygromycin B) are MMLV particles designed to transduce adherent cells. This allows production of PL-tagged, hygromycin B-resistant target cells of interest for use in cytotoxicity assays involving detection of cell lysis. This product uses EFC technology. The retroparticle mix can be used to transduce target cells with a PL-tagged housekeeping gene. Target cell death in a cytotoxicity assay will release PL to the media, where EA contained in the detection reagent will produce EFC signal. These retroparticles ship as 4 vials of cryopreserved retroparticles.
Specifications
For Use With (Application) Retroparticles
Product Type Retroparticles
Quantity 4 Vials