Learn More
DWK Life Sciences KIMBLE™ Glass Screw-Thread Sample Vials with PTFE Closure
Shop All DWK Life Sciences ProductsDescription
Cylindrical shape provides solid balance. Flat bottom facilitates free-standing capability.
- Clear borosilicate glass sample vials with cylindrical shape and flat bottom for free-standing stability
- Screw-thread GPI finish for secure, leak-resistant closure
- Supplied with attached closed-top white urea cap with PTFE-faced white rubber liner
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
- 60940A-4 complies with EPA 600 series methods
- Packaged in corrugated trays with partitions
Specifications
Specifications
| Classification | A |
| Color | Clear |
| Diameter (Metric) Outer | 21 mm |
| Material | Borosilicate Glass |
| Pre-slit | No |
| Sterility | Non-sterile |
| Volume (Metric) | 16.0 mL |
| Thread Style | Screw |
| Volume (English) | 4.32 dr. |
| Closure Material | PTFE |
| Show More |