Learn More
Thermo Scientific™ Kinase Buffer R (5X)
Product Code. 10528170
Change view
Click to view available options
Quantity:
4 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10528170 4 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10528170 Supplier Thermo Scientific™ Supplier No. PV5101
 Restricted Item
Expédition estimée: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Kinase buffer that has been specially optimized for use with the Class I & II PI3 kinases.
Kinase buffer that has been specially optimized for use with the Class I & II PI3 kinases.
TRUSTED_SUSTAINABILITY
Specifications
Concentration 5X
For Use With (Equipment) Pharma and biopharma, protein biology, drug discovery and development, target and lead identification and validation, kinase biology, biochemical kinase assays, Lanthascreen™ kinase activity assays, Adapta™ Universal kinase assays, enzyme and
Physical Form Liquid
Quantity 4 mL
Form Liquid
For Research Use Only. Not for use in diagnostic procedures.