Learn More
Thermo Scientific™ Kinase Buffer T (5X)
Product Code. 10092803
Change view
Click to view available options
Quantity:
4 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10092803 4 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10092803 Supplier Thermo Scientific™ Supplier No. PV5373
 Restricted Item
Expédition estimée: 02-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Kinase buffer that has been specially optimized for use with the Type III PI4 kinases including PI4KA and PI4KB.
Kinase buffer that has been specially optimized for use with the Type III PI4 kinases including PI4KA and PI4KB.
TRUSTED_SUSTAINABILITY
Specifications
Concentration 5X
For Use With (Equipment) Pharma and biopharma, target and lead identification and validation
Packaging 1 Each
Physical Form Liquid
Quantity 4 mL
Form Liquid
For Research Use Only. Not for use in diagnostic procedures.